Läs mer
Dihydro Finasteride, TRC
Produktkod. 17919639
Change view
Kvantitet:
25 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17919639 25 mg
18073430 500 mg
18010430 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17919639 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44897525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 98319-24-5
Molekylformel C23 H38 N2 O2
Molekylvikt (g/mol) 374.56
Synonym N-(1,1-Dimethylethyl)-3-oxo-4-aza-5alpha-androstane-17beta-carboxamide, Finasteride Imp. A (EP), Dihydrofinasteride, 1H-Indeno[5,4-f]quinoline-7-carboxamide, N-(1,1-dimethylethyl)hexadecahydro-4a,6a-dimethyl-2-oxo-, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-, 4-Azaandrostane-17-carboxamide, N-(1,1-dimethylethyl)-3-oxo-, (5α,17β)-, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-N-(1,1-Dimethylethyl)hexadecahydro-4a,6a-dimethyl-2-oxo-1H-indeno[5,4-f]quinoline-7-carboxamide, 1,2-Dihydrofinasteride, 1H-Indeno[5,4-f]quinoline-7-carboxamide, N-(1,1-dimethylethyl)hexadecahydro-4a,6a-dimethyl-2-oxo-, [4aR-(4aα,4bβ,6aα,7α,9aβ,9bα,11aβ)]-, 3-Oxo-4-aza-5α-androstan-17β-carboxylic acid N-(tert-butyl)amide, N-(tert-Butyl)-3-oxo-4-aza-5α-androstane-17-carboxamide, N-(tert-Butyl)-3-oxo-4-aza-5α-androstane-17β-carboxamide
IUPAC-namn (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,8,9,9b,10,11-tetradecahydroindeno[5,4-f]quinoline-1-carboxamide
LEDER CC(C)(C)NC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4NC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material Dihydrofinasteride
Kvantitet 25 mg
InChI formel InChI=1S/C23H38N2O2/c1-21(2,3)25-20(27)17-8-7-15-14-6-9-18-23(5,13-11-19(26)24-18)16(14)10-12-22(15,17)4/h14-18H,6-13H2,1-5H3,(H,24,26)(H,25,27)/t14-,15-,16-,17+,18+,22-,23+/m0/s1
Formel vikt 374.2933