Läs mer
Dihydro Ketoprofen (Mixture of Diastereomers), TRC
Produktkod. 30009665
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30009665 2.5 mg 2.5mg
30008046 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009665 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4491762.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 59960-32-6
Molekylformel C16 H16 O3
Molekylvikt (g/mol) 256.3
Synonym 2-[3-(alpha-Hydroxybenzyl)phenyl]propanoic Acid
IUPAC-namn 2-[3-[hydroxy(phenyl)methyl]phenyl]propanoic acid
LEDER CC(C(=O)O)c1cccc(c1)C(O)c2ccccc2
Specifikationer
Kemiskt namn eller material 2-[3-(alpha-Hydroxybenzyl)phenyl]propanoic Acid
Kvantitet 2.5 mg
InChI formel InChI=1S/C16H16O3/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-11,15,17H,1H3,(H,18,19)
Formel vikt 256.1099