Läs mer
Dihydro Montelukast, TRC
Produktkod. 30011000
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011000 1 mg
30004786 5 mg
30006460 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011000 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD5492151MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 142147-67-9
Molekylformel C35 H38 Cl N O3 S
Molekylvikt (g/mol) 588.2
Synonym 1-[[[(1R)-1-[3-[2-(7-Chloro-2-quinolinyl)ethyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]cyclopropaneacetic Acid, Cyclopropaneacetic acid, 1-[[[1-[3-[2-(7-chloro-2-quinolinyl)ethyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]-, (R)- (9CI), 1-[[[(1R)-1-[3-[2-(7-Chloro-2-quinolinyl)ethyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]cyclopropaneacetic acid
IUPAC-namn 2-[1-[[(1R)-1-[3-[2-(7-chloroquinolin-2-yl)ethyl]phenyl]-3-[2-(2-hydroxypropan-2-yl)phenyl]propyl]sulfanylmethyl]cyclopropyl]acetic acid
LEDER CC(C)(O)c1ccccc1CC[C@@H](SCC2(CC(=O)O)CC2)c3cccc(CCc4ccc5ccc(Cl)cc5n4)c3
Specifikationer
Kemiskt namn eller material Dihydro Montelukast
Kvantitet 1 mg
InChI formel InChI=1S/C35H38ClNO3S/c1-34(2,40)30-9-4-3-7-25(30)13-17-32(41-23-35(18-19-35)22-33(38)39)27-8-5-6-24(20-27)10-15-29-16-12-26-11-14-28(36)21-31(26)37-29/h3-9,11-12,14,16,20-21,32,40H,10,13,15,17-19,22-23H2,1-2H3,(H,38,39)/t32-/m1/s1
Formel vikt 587.2261