Läs mer
Dihydroabietic Acid (Technical Grade), TRC
Produktkod. 30369154
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30369154 25 mg
1 options
Produktkod. 30369154 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD48793825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dihydroabietic Acid
CAS 2221-96-7
Molekylformel C20 H32 O2
Kvantitet 25 mg
Synonym A-Dihydroabietic Acid, 13α-Isopropyl-podocarp-8(14)-en-15-oic Acid, (1R,4aR,4bS,7R,10aR)-1,2,3,4,4a,4b,5,6,7,9,10,10a-Dodecahydro-1,4a-dimethyl-7-(1-methylethyl)-1-phenanthrenecarboxylic Acid, 13βH-δ8(14)-Dihydroabietic Acid, 13βH-δ8(14)-dihydroabietic Acid
InChI formel InChI=1S/C20H32O2/c1-13(2)14-6-8-16-15(12-14)7-9-17-19(16,3)10-5-11-20(17,4)18(21)22/h12-14,16-17H,5-11H2,1-4H3,(H,21,22)/t14-,16-,17+,19+,20+/m0/s1
LEDER CC(C)[C@H]1CC[C@H]2C(=C1)CC[C@@H]3[C@]2(C)CCC[C@@]3(C)C(=O)O
IUPAC-namn (1R,4aR,4bS,7R,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,7,9,10,10a-decahydrophenanthrene-1-carboxylic acid
Molekylvikt (g/mol) 304.467
Formel vikt 304.24