Läs mer
Dihydroartemisinic Acid, TRC
Produktkod. 30134800
Change view
Kvantitet:
10 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134800 1 g
30134621 250 mg
30011959 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134800 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4485181G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 85031-59-0
Molekylformel C15H24O2
Molekylvikt (g/mol) 236.35
Synonym (αR,1R,4R,4aS,8aS)-1,2,3,4,4a,5,6,8a-Octahydro-α,4,7-trimethyl-1-naphthaleneacetic Acid, Dihydroarteannuic Acid, Dihydroqinghao Acid, [1R-[1α(R*),4β,4aβ,8aβ]]-1,2,3,4,4a,5,6,8a-Octahydro-α,4,7-trimethyl-1-naphthaleneacetic Acid;
IUPAC-namn (2R)-2-[(1R,4R,4aS,8aS)-4,7-dimethyl-1,2,3,4,4a,5,6,8a-octahydronaphthalen-1-yl]propanoic acid
LEDER CC1=C[C@H]2[C@@H](CC1)[C@H](C)CC[C@H]2[C@@H](C)C(=O)O
Specifikationer
Kemiskt namn eller material Dihydroartemisinic Acid
Kvantitet 1 g
InChI formel InChI=1S/C15H24O2/c1-9-4-6-12-10(2)5-7-13(14(12)8-9)11(3)15(16)17/h8,10-14H,4-7H2,1-3H3,(H,16,17)/t10-,11-,12+,13+,14+/m1/s1
Formel vikt 236.1776