Läs mer
Dihydroartemisinin, MedChemExpress
Produktkod. 16489016
Encompass_Preferred
Change view
Kvantitet:
50 mg
100 mg
200 mg
500 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16489016 100 mg
16492356 500 mg
16474777 50 mg
16461496 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16489016 Leverantör Medchem Express Leverantörsnummer HYN0176
MedChemExpress Dihydroartemisinin is a potent anti-malaria agent.
Dihydroartemisinin is a potent anti-malaria agent.
Specifikationer
Kemiskt namn eller material Dihydroartemisinin
Fysisk form Solid
CAS 71939-50-9
Färg White
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C15H24O5
För användning med (applikation) Cancer-programmed cell death
Synonym Dihydroqinghaosu β-Dihydroartemisinin Artenimol
Löslighetsinformation DMSO : 25 mg/mL (87.92 mM; Need ultrasonic) ∣Ethanol : 10 mg/mL (35.17 mM; Need ultrasonic)
LEDER O[C@@H]1[C@H](C)[C@]2([H])CC[C@@H](C)[C@]3([H])CC[C@@](O4)(C)OO[C@]32[C@]4([H])O1
Molekylvikt (g/mol) 284.35
Formel vikt 284.35
Procent renhet 98.0%