Läs mer
Dihydrocapsaicin, LoGiCal
Produktkod. 30460839
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30460839 10 mg
1 options
Produktkod. 30460839 Leverantör LoGiCal Leverantörsnummer LGCFOR0016.01
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dihydrocapsaicin
CAS 19408-84-5
Analyt- eller komponentnamn Dihydrocapsaicin
Frakt skick Room Temperature
Molekylformel C18 H29 N O3
Certifikat och efterlevnad ISO 9001
Synonym Dihydrocapsaicin, Nonanamide, 8-methyl-N-vanillyl- (7CI,8CI), 6,7-Dihydrocapsaicin, Capsaicin, dihydro-, Dihydrocapsaicin
InChI formel InChI=1S/C18H29NO3/c1-14(2)8-6-4-5-7-9-18(21)19-13-15-10-11-16(20)17(12-15)22-3/h10-12,14,20H,4-9,13H2,1-3H3,(H,19,21)
Lösningstyp Neat
LEDER COc1cc(CNC(=O)CCCCCCC(C)C)ccc1O
IUPAC-namn N-[(4-hydroxy-3-methoxyphenyl)methyl]-8-methylnonanamide
Industry Type Clinical Testing
Molekylvikt (g/mol) 307.43
Kvantitet 10 mg
Formel vikt 307.2147 g/mol