Läs mer
Dihydrocurcumin, TRC
Produktkod. 30378163
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30378163 5 mg
1 options
Produktkod. 30378163 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4518005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dihydrocurcumin
CAS 943330-36-7
Molekylformel C21H22O6
Kvantitet 5 mg
Synonym (1E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)-1-Heptene-3,5-dione
InChI formel InChI=1S/C21H22O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)13-17(23)8-4-15-6-10-19(25)21(12-15)27-2/h3,5-7,9-12,24-25H,4,8,13H2,1-2H3/b7-3+
LEDER COc1cc(CCC(=O)CC(=O)\C=C\c2ccc(O)c(OC)c2)ccc1O
Molekylvikt (g/mol) 370.4
Formel vikt 370.1416