missing translation for 'onlineSavingsMsg'
Learn More

Dihydrogen hexafluorozirconate, 45% w/w aq. soln., Thermo Scientific Chemicals

Product Code. p-7050792
500 g, 500g
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
This item is not returnable. View return policy
This item is not returnable. View return policy

Dihydrogen hexafluorozirconate is used in manufacturing of optical glass and fluozirconate. It acts as a precursor to zirconium dioxide ceramic thin films. Further, it is used for surface pretreatments on steel to prevent corrosion. In addition to this, it is used in the production of the atomic energy industry, advanced electrical materials and refractories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemische Identifikatoren

CAS 12021-95-3
Molecular Formula F6H2Zr
Molecular Weight (g/mol) 207.23
MDL Number MFCD00082965
InChI Key PZFQYBOZXGWNAM-UHFFFAOYSA-H
Synonym Hexafluorozirconic acid; Hydrogen hexafluorozirconate

Spezifikation

Color Colorless
Density 1.51g/mL
Quantity 500 g
UN Number UN2922
Formula Weight 207.22
Concentration or Composition (by Analyte or Components) 45% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Dihydrogen hexafluorozirconate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberS03L002 Certificate TypeCertificate of Analysis Date03/06/2025 Product Code039462.36
Lot NumberX26J025 Certificate TypeCertificate of Analysis Date03/11/2023 Product Code039462.A3,039462.36
Lot NumberU07G029 Certificate TypeCertificate of Analysis Date11/10/2023 Product Code039462.A3,039462.36
Lot NumberR14F012 Certificate TypeCertificate of Analysis Date06/10/2023 Product Code039462.A3,039462.36
Lot NumberZ20D021 Certificate TypeCertificate of Analysis Date04/10/2023 Product Code039462.A3,039462.36

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Dihydrogen hexafluorozirconate, 45% w/w aq. soln., Thermo Scientific Chemicals >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado