Läs mer
Dihydroisolysergic Acid Methyl Ester, TRC
Produktkod. 30376918
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30376918 1 mg
1 options
Produktkod. 30376918 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4502551MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dihydroisolysergic Acid Methyl Ester
CAS 5143-94-2
Molekylformel C17H20N2O2
Kvantitet 1 mg
Synonym 9,10-Dihydroisolysergic Acid I Methyl Ester, 6-Methylergoline-8-carboxylic Acid Methyl Ester
InChI formel InChI=1S/C17H20N2O2/c1-19-9-11(17(20)21-2)6-13-12-4-3-5-14-16(12)10(8-18-14)7-15(13)19/h3-5,8,11,13,15,18H,6-7,9H2,1-2H3/t11-,13+,15+/m0/s1
LEDER [H][C@]1([C@]2([H])C[C@H](C(OC)=O)CN1C)CC3=CNC4=C3C2=CC=C4
IUPAC-namn methyl (6aR,9S,10aR)-7-methyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinoline-9-carboxylate
Molekylvikt (g/mol) 284.35
Formel vikt 284.15