Läs mer
Dihydrolysergic Acid Methyl Ester, TRC
Produktkod. 30372320
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30372320 10 mg
1 options
Produktkod. 30372320 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45026510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dihydrolysergic Acid Methyl Ester
CAS 35470-53-2
Molekylformel C17H20N2O2
Kvantitet 10 mg
Synonym Methyl 6-Methylergoline-8beta-carboxylate, Methyl 9,10-Dihydro-D-lysergate, Methyl 9,10-Dihydrolysergate, Methyl D-6-Methylergoline-8beta-carboxylate, Methyl Dihydrolysergate, 9,10-Dihydrolysergic Acid Methyl EsterD-Dihydrolysergic Acid Methyl Ester
InChI formel InChI=1S/C17H20N2O2/c1-19-9-11(17(20)21-2)6-13-12-4-3-5-14-16(12)10(8-18-14)7-15(13)19/h3-5,8,11,13,15,18H,6-7,9H2,1-2H3/t11-,13-,15-/m1/s1
LEDER COC(=O)[C@@H]1C[C@H]2[C@@H](Cc3c[nH]c4cccc2c34)N(C)C1
IUPAC-namn methyl (6aR,9R,10aR)-7-methyl-6,6a,8,9,10,10a-hexahydro-4H-indolo[4,3-fg]quinoline-9-carboxylate
Molekylvikt (g/mol) 284.35
Formel vikt 284.1525