Läs mer
Dihydroorotate Acid Methyl Ester, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dihydroorotate Acid Methyl Ester |
| CAS | 23903-57-3 |
| Molekylformel | C6 H8 N2 O4 |
| Kvantitet | 50 mg |
| Synonym | Hydroorotic Acid Methyl Ester, Methyl Dihydroorotate, Hexahydro-2,6-dioxo-4-pyrimidinecarboxylic Acid Methyl Ester |
| InChI formel | InChI=1S/C6H8N2O4/c1-12-5(10)3-2-4(9)8-6(11)7-3/h3H,2H2,1H3,(H2,7,8,9,11) |
| LEDER | COC(=O)C1CC(=O)NC(=O)N1 |
| IUPAC-namn | methyl 2,6-dioxo-1,3-diazinane-4-carboxylate |
| Molekylvikt (g/mol) | 172.14 |
| Formel vikt | 172.0484 |