Läs mer
(+)-Dihydroquinidine, TRC
Produktkod. 30377561
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30377561 1 g
1 options
Produktkod. 30377561 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4520951G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (+)-Dihydroquinidine
CAS 1435-55-8
Molekylformel C20H26N2O2
Kvantitet 1 g
Synonym Cinchonan-9-ol, 10,11-dihydro-6'-methoxy-, (9S)-, Hydroquinidine (6CI,7CI,8CI), (+)-Hydroquinidine, (9S)-10,11-Dihydro-6'-methoxycinchonan-9-ol, 10,11-Dihydroquinidine, Dihydroquinidine, Hydroconquinine
InChI formel InChI=1S/C20H26N2O2/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18/h4-6,8,11,13-14,19-20,23H,3,7,9-10,12H2,1-2H3/t13-,14-,19+,20-/m0/s1
LEDER CC[C@H]1CN2CC[C@H]1C[C@@H]2[C@@H](O)c3ccnc4ccc(OC)cc34
Molekylvikt (g/mol) 326.43
Formel vikt 326.1994