Läs mer
Dihydroxy Moxonidine, TRC
Produktkod. 30377799
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30377799 2.5 mg 2.5mg
30377446 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30377799 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4534602.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 352457-32-0
Molekylformel C8H11N5O2
Molekylvikt (g/mol) 209.21
Synonym 5-[(4,5-Dihydro-1H-imidazol-2-yl)amino]-6-hydroxy-2-methyl-4(3H)-pyrimidinone, Moxonidine Impurity 1
LEDER Cc1nc(O)c(N=C2NCCN2)c(O)n1
Specifikationer
Kemiskt namn eller material Dihydroxy Moxonidine
Kvantitet 2.5 mg
InChI formel InChI=1S/C8H11N5O2/c1-4-11-6(14)5(7(15)12-4)13-8-9-2-3-10-8/h2-3H2,1H3,(H2,9,10,13)(H2,11,12,14,15)
Formel vikt 209.0913