Läs mer
Dihydroxy Oxaliplatin-Pt(IV), TRC
Produktkod. 30137620
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137620 5 mg
30012854 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137620 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4541855MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 111321-67-6
Molekylformel C8 H16 N2 O6 Pt
Molekylvikt (g/mol) 431.3
Synonym Platinum, [(1R,2R)-1,2-cyclohexanediamine-kappaN1,kappaN2][ethanedioato(2-)-kappaO1,kappaO2]dihydroxy-, (OC-6-33)-, Platinum, (1,2-cyclohexanediamine-N,N')[ethanedioato(2-)-O,O']dihydroxy-, [OC-6-33-(1R-trans)]- (9CI), 1,2-Cyclohexanediamine, platinum complex, (1R-trans)-, Oxaliplatin Imp. C (EP), Oxaliplatin Related Compound C (USP)
LEDER O[Pt+2]12(O)([NH2][C@@H]3CCCC[C@H]3[NH2]1)[O-]C(=O)C(=O)[O-]2
Specifikationer
Kemiskt namn eller material (OC-6-33)-[(1R,2R)-Cyclohexane-1,2-diamine-kappaN,kappaN'][ethanedioato(2-)-kappaO1,kappaO2]-dihydroxyplatinum
Kvantitet 5 mg
InChI formel InChI=1S/C6H14N2.C2H2O4.2H2O.Pt/c7-5-3-1-2-4-6(5)8;3-1(4)2(5)6;;;/h5-6H,1-4,7-8H2;(H,3,4)(H,5,6);2*1H2;/q;;;;+4/p-4/t5-,6-;;;;/m1..../s1
Formel vikt 431.0656