Läs mer
Dihydroxybis(2-methyl-2-phenylpropyl)-stannane (Technical Grade), TRC
Produktkod. 30428413
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428413 25 mg
30414484 100 mg
30415986 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30428413 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS12540025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 162476-39-3
Molekylformel C20 H28 O2 Sn
Molekylvikt (g/mol) 419.15
Synonym Stannane, dihydroxybis(2-methyl-2-phenylpropyl)-, Dihydroxybis(2-methyl-2-phenylpropyl)stannane
IUPAC-namn dihydroxy-bis(2-methyl-2-phenylpropyl)stannane
LEDER CC(C)(C[Sn](O)(O)CC(C)(C)c1ccccc1)c2ccccc2
Specifikationer
Kemiskt namn eller material Dihydroxy-bis(2-methyl-2-phenylpropyl)stannane
Kvantitet 25 mg
InChI formel InChI=1S/2C10H13.2H2O.Sn/c2*1-10(2,3)9-7-5-4-6-8-9;;;/h2*4-8H,1H2,2-3H3;2*1H2;/q;;;;+2/p-2
Formel vikt 420.1111