missing translation for 'onlineSavingsMsg'
Läs mer

Dihydrozeatin Riboside (>90%), TRC

Product Code. 30007914
Change view
Click to view available options
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30007914 5 mg
30011255 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007914 Supplier Toronto Research Chemicals Supplier No. TRCD4548405MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22663-55-4
Molekylformel C15H23N5O5
Molekylvikt (g/mol) 353.37
Synonym 6-(4-Hydroxy-3-methylbutylamino)-9-ß-dribofuranosylpurine, Ribosyldihydrozeatin, Dihydrozeatin 9-Riboside, Dihydrozeatin Ribonucleoside, 9-beta-D-Ribofuranosyldihydrozeatin, N-(4-Hydroxy-3-methylbutyl)adenosine
IUPAC-namn (2R,3R,4S,5R)-2-(hydroxymethyl)-5-[6-[(4-hydroxy-3-methylbutyl)amino]purin-9-yl]oxolane-3,4-diol
LEDER CC(CO)CCNc1ncnc2c1ncn2[C@@H]3O[C@H](CO)[C@H](O)[C@@H]3O

Specifications

Kemiskt namn eller material Dihydrozeatin Riboside
Kvantitet 5 mg
InChI formel InChI=1S/C15H23N5O5/c1-8(4-21)2-3-16-13-10-14(18-6-17-13)20(7-19-10)15-12(24)11(23)9(5-22)25-15/h6-9,11-12,15,21-24H,2-5H2,1H3,(H,16,17,18)/t8?,9-,11+,12+,15-/m1/s1
Formel vikt 353.1699
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.