missing translation for 'onlineSavingsMsg'
Läs mer

Dihydrozeatin Riboside (>90%), TRC

Produktkod. 30011255
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011255 10 mg
30007914 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30011255 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45484010MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22663-55-4
Molekylformel C15H23N5O5
Molekylvikt (g/mol) 353.37
Synonym 6-(4-Hydroxy-3-methylbutylamino)-9-ß-dribofuranosylpurine, Ribosyldihydrozeatin, Dihydrozeatin 9-Riboside, Dihydrozeatin Ribonucleoside, 9-beta-D-Ribofuranosyldihydrozeatin, N-(4-Hydroxy-3-methylbutyl)adenosine
IUPAC-namn (2R,3R,4S,5R)-2-(hydroxymethyl)-5-[6-[(4-hydroxy-3-methylbutyl)amino]purin-9-yl]oxolane-3,4-diol
LEDER CC(CO)CCNc1ncnc2c1ncn2[C@@H]3O[C@H](CO)[C@H](O)[C@@H]3O

Specifications

Kemiskt namn eller material Dihydrozeatin Riboside
Kvantitet 10 mg
InChI formel InChI=1S/C15H23N5O5/c1-8(4-21)2-3-16-13-10-14(18-6-17-13)20(7-19-10)15-12(24)11(23)9(5-22)25-15/h6-9,11-12,15,21-24H,2-5H2,1H3,(H,16,17,18)/t8?,9-,11+,12+,15-/m1/s1
Formel vikt 353.1699
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.