missing translation for 'onlineSavingsMsg'
Learn More

Diiron nonacarbonyl, 98%, Thermo Scientific Chemicals

Catalog Number p-7203424
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
15321-51-4
C9H3Fe2O9
MFCD00151465
CGJSNKHFIBINMR-UHFFFAOYSA-N
iron nonacarbonyl, enneacarbonyldiiron, nonacarbonyldiiron, 1,3-diferrabicyclo 1.1.1 pentane-2,4,5-trione; hexakis carbon monoxide, diironnonacarbonyl, diironnonacarbonyl, purum fe
71311424
This item is not returnable. View return policy
15321-51-4
C9H3Fe2O9
MFCD00151465
CGJSNKHFIBINMR-UHFFFAOYSA-N
iron nonacarbonyl, enneacarbonyldiiron, nonacarbonyldiiron, 1,3-diferrabicyclo 1.1.1 pentane-2,4,5-trione; hexakis carbon monoxide, diironnonacarbonyl, diironnonacarbonyl, purum fe
71311424

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Black to Brown-Red
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle packed in tin can
Linear Formula Fe2(CO)9
Quantity 10 g
Fieser 01,259; 02,139; 03,101; 04,157; 05,221; 06,195; 07,110; 08,498; 13,320; 04,534; 05,706; 07,144; 08,41; 08,216; 10,174; 08,214
SMILES [CH-]=O.[CH-]=O.[CH-]=O.[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Fe].[Fe+3]
IUPAC Name carbon monoxide;iron;iron(3+);methanone
Molecular Weight (g/mol) 366.804
Formula Weight 363.78
Percent Purity ≥97.5%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Diiron nonacarbonyl, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Diiron nonacarbonyl
Signal Word
  • Danger
Hazard Category
  • Pyrophoric solid Category 1
Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P243-Take precautionary measures against static discharge.
  • P422-Store contents under .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'