Läs mer
Diisoheptyl Phthalate (mixture of C7 isomers), TRC
Produktkod. 30378541
Change view
Kvantitet:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30378541 5 mL
30371708 100 mL
30372156 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30378541 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4557205ML
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 71888-89-6
Molekylformel C22H34O4
Molekylvikt (g/mol) 362.5
Synonym Phthalic acid, bis-C6-C8-branched alkyl esters C7-rich
IUPAC-namn diheptyl phthalate
LEDER O=C(OCCCCCCC)C1=CC=CC=C1C(OCCCCCCC)=O
Specifikationer
Kemiskt namn eller material Phthalic acid, bis-C6-C8-branched alkyl esters C7-rich
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mL
InChI formel InChI=1S/C22H34O4/c1-3-5-7-9-13-17-25-21(23)19-15-11-12-16-20(19)22(24)26-18-14-10-8-6-4-2/h11-12,15-16H,3-10,13-14,17-18H2,1-2H3
Formel vikt 222.0892
Procent renhet >95%