Läs mer
Diisononyl Phthalate, TRC
Produktkod. 30008854
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008854 100 mg
30005813 50 mg
17902075 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008854 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD455225100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 20548-62-3
Molekylformel C26 H42 O4
Molekylvikt (g/mol) 418.6093
Synonym 1,2-Benzenedicarboxylic acid, bis(7-methyloctyl) ester (9CI), Phthalic acid, bis(7-methyloctyl) ester (7CI,8CI), 1-Octanol, 7-methyl-, phthalate (2:1) (8CI), Bis(7-methyloctyl) phthalate, Diisononyl phthalate
IUPAC-namn bis(7-methyloctyl) benzene-1,2-dicarboxylate
LEDER CC(C)CCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCC(C)C
Specifikationer
Kemiskt namn eller material Phthalic acid, bis-7-methyloctyl ester
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C26H42O4/c1-21(2)15-9-5-7-13-19-29-25(27)23-17-11-12-18-24(23)26(28)30-20-14-8-6-10-16-22(3)4/h11-12,17-18,21-22H,5-10,13-16,19-20H2,1-4H3
Formel vikt 418.3083
Procent renhet >95%