Läs mer
Diisopentyl Phthalate, TRC
Produktkod. 30179632
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179632 1 g
30179292 250 mg
30179095 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179632 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4552301G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 605-50-5
Molekylformel C18 H26 O4
Molekylvikt (g/mol) 306.4
Synonym Phthalic Acid, Bis-isopentyl Ester, 1,2-Benzenedicarboxylic acid, 1,2-bis(3-methylbutyl) ester (ACI), 1,2-Benzenedicarboxylic acid, bis(3-methylbutyl) ester (9CI), Phthalic acid, diisopentyl ester (6CI, 7CI, 8CI), Bis(3-methylbutyl) phthalate, Diisoamyl phthalate, Diisopentyl phthalate, NSC 17070, Palatinol DIPP
IUPAC-namn bis(3-methylbutyl) benzene-1,2-dicarboxylate
LEDER CC(C)CCOC(=O)c1ccccc1C(=O)OCCC(C)C
Specifikationer
Kemiskt namn eller material Phthalic acid, bis-iso-pentyl ester
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C18H26O4/c1-13(2)9-11-21-17(19)15-7-5-6-8-16(15)18(20)22-12-10-14(3)4/h5-8,13-14H,9-12H2,1-4H3
Formel vikt 306.1831
Procent renhet >95%