missing translation for 'onlineSavingsMsg'
Learn More

(+)-Diisopinocampheylchloroborane, 1.8M solution in hexanes, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-3586320
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
112246-73-8, 80-56-8, 71-43-2, 64742-89-8
C20H34BCl
320.75
MFCD00074808
PSEHHVRCDVOTID-NAVXHOJHSA-N
+-b-chlorodiisopinocamphenylborane
133124869
chloro-bis[(3R,4S,5R)-4,6,6-trimethyl-3-bicyclo[3.1.1]heptanyl]borane
C[C@H]1[C@@H]2C[C@H](C[C@@H]1B(Cl)[C@H]1C[C@H]3C[C@@H]([C@@H]1C)C3(C)C)C2(C)C
This item is not returnable. View return policy
112246-73-8, 80-56-8, 71-43-2, 64742-89-8
C20H34BCl
320.75
MFCD00074808
PSEHHVRCDVOTID-NAVXHOJHSA-N
+-b-chlorodiisopinocamphenylborane
133124869
chloro-bis[(3R,4S,5R)-4,6,6-trimethyl-3-bicyclo[3.1.1]heptanyl]borane
C[C@H]1[C@@H]2C[C@H](C[C@@H]1B(Cl)[C@H]1C[C@H]3C[C@@H]([C@@H]1C)C3(C)C)C2(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Solubility Information Solubility in water: reacts
Formula Weight 320.77
Chemical Name or Material (+)-Diisopinocampheylchloroborane
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'