Läs mer
Diisopropyl Adipate, TRC
Produktkod. 17987454
Change view
Kvantitet:
5 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17987454 5 mL
17968684 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17987454 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4598605ML
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6938-94-9
Molekylformel C12 H22 O4
Molekylvikt (g/mol) 230.3
Synonym Ceraphyl 230, Crodamol DA, Diisopropyl Adipate, Iso Adipat, Isopropyl Adipate, NSC 56587, Nikkol DID, Schercemol DIA, Standamul DIPA, Wickenol 116;
IUPAC-namn diisopropyl hexanedioate
LEDER CC(C)OC(=O)CCCCC(=O)OC(C)C
Specifikationer
Kemiskt namn eller material Diisopropyl Adipate
Kvantitet 5 mL
InChI formel InChI=1S/C12H22O4/c1-9(2)15-11(13)7-5-6-8-12(14)16-10(3)4/h9-10H,5-8H2,1-4H3
Formel vikt 230.1518