missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diisopropyl ether, puriss p.a., ≥98.5% (GC), Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Melting Point | -86°C |
Density | 0.724g/mL at 20°C, 0.725g/mL at 25°C |
Boiling Point | 67°C to 70°C |
Flash Point | −29°C (−20.2°F) |
Packaging | Glass bottle |
Linear Formula | (CH3)2CHOCH(CH3)2 |
Refractive Index | 1.367 |
Quantity | 1 L |
UN Number | UN1159 |
Beilstein | 1731256 |
Show More |
Safety and Handling
Product Identifier
- Diisopropyl ether
- Danger
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'