Läs mer
Diisopropyl Fluorophosphate, TRC
Produktkod. 30132631
Change view
Kvantitet:
1 g
3 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132631 3 g
30040488 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132631 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4553003G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55-91-4
Molekylformel C6 H14 F O3 P
Molekylvikt (g/mol) 184.15
Synonym Phosphorofluoridic acid, bis(1-methylethyl) ester, Phosphorofluoridic acid, diisopropyl ester (8CI), DFF, DFP, DFP (pesticide), Difluorophate, Diflupyl, Diisopropoxyphosphoryl fluoride, Diisopropyl fluorophosphate, Diisopropyl phosphofluoridate, Diisopropyl phosphorofluoridate, Dyflos, Floropryl, Fluorodiisopropyl phosphate, Fluostigmine, Isoflurophate, Isopropyl fluophosphate ((C3H7O)2FPO), Isopropyl phosphorofluoridate ((C3H7O)2FPO), Neoglaucit, O,O-Diisopropyl phosphorofluoridate
IUPAC-namn 2-[fluoro(propan-2-yloxy)phosphoryl]oxypropane
LEDER CC(C)OP(=O)(F)OC(C)C
Specifikationer
Kemiskt namn eller material Diisopropyl Fluorophosphate
Kvantitet 3 g
InChI formel InChI=1S/C6H14FO3P/c1-5(2)9-11(7,8)10-6(3)4/h5-6H,1-4H3
Formel vikt 184.0665