Learn More
Diisopropyl Hydrazine-1,2-dicarboxylate, Mikromol™
Product Code. 30051709
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051709 25 mg 25mg
30049783 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30051709 Supplier Mikromol Supplier No. MM1219.090025
 Restricted Item
Beräknad leverans: 27-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Diisopropyl Hydrazine-1,2-dicarboxylate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Diisopropyl Bicarbamate
CAS 19740-72-8
Analyte or Component Names Diisopropyl Bicarbamate
Shipping Condition Room Temperature
Molecular Formula C8 H16 N2 O4
Certifications/Compliance ISO 17025
Synonym Diisopropyl bicarbamate, Diisopropyl Hydrazine-1,2-dicarboxylate, Orlistat Related Compound B
InChI Formula InChI=1S/C8H16N2O4/c1-5(2)13-7(11)9-10-8(12)14-6(3)4/h5-6H,1-4H3,(H,9,11)(H,10,12)
SMILES CC(C)OC(=O)NNC(=O)OC(C)C
IUPAC Name propan-2-yl N-(propan-2-yloxycarbonylamino)carbamate
Molecular Weight (g/mol) 204.22
Quantity 25 mg
Formula Weight 204.111 g/mol