Läs mer
Diisopropyl Sebacate, TRC
Produktkod. 30373464
Change view
Kvantitet:
2.5 g
Produktkod. Kvantitet
30373464 2.5 g
1 options
Produktkod. 30373464 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD5257932.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diisopropyl Sebacate
CAS 7491-02-3
Molekylformel C16H30O4
Kvantitet 2.5 g
Synonym Sebacic Acid, Bis-isopropyl Ester, Decanedioic acid, bis(1-methylethyl) ester (9CI), Sebacic acid, diisopropyl ester (6CI, 8CI), Dermol DIPS, Diisopropyl sebacate, IPSE, Schercemol DIS, Wickenol 117
InChI formel InChI=1S/C16H30O4/c1-13(2)19-15(17)11-9-7-5-6-8-10-12-16(18)20-14(3)4/h13-14H,5-12H2,1-4H3
LEDER CC(C)OC(=O)CCCCCCCCC(=O)OC(C)C
IUPAC-namn dipropan-2-yl decanedioate
Molekylvikt (g/mol) 286.41
Formel vikt 286.2144