Läs mer
Diisopropylformamide Dimethylacetal (>90%), TRC
Produktkod. 30372639
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30372639 25 mg
1 options
Produktkod. 30372639 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45539825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Diisopropylformamide Dimethylacetal
CAS 19449-28-6
Molekylformel C9H21NO2
Kvantitet 25 mg
Synonym N-(Dimethoxymethyl)-N-(1-methylethyl)-2-propanamine, N,N-Diisopropylformamide Dimethyl Acetal, N-(Dimethoxymethyl)-1,1'-dimethyl-diethylamine
InChI formel InChI=1S/C9H21NO2/c1-7(2)10(8(3)4)9(11-5)12-6/h7-9H,1-6H3
LEDER COC(OC)N(C(C)C)C(C)C
IUPAC-namn N-(dimethoxymethyl)-N-propan-2-ylpropan-2-amine
Molekylvikt (g/mol) 175.27
Formel vikt 175.1572