Läs mer
Diisoundecyl Phthalate (Mixture of Branched Isomers), TRC
Produktkod. 17999084
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17999084 1 g
17987544 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17999084 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4599001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 96507-86-7
Molekylformel C30 H50 O4
Molekylvikt (g/mol) 474.716
Synonym 1,2-Benzenedicarboxylic acid, diisoundecyl ester (9CI), Diisoundecyl phthalate, DIUP, Jayflex DIUP, Phthalic Acid, Bis-Isoundecyl Ester (Mixture Of Branched Chain Isomers), 1,2-Benzenedicarboxylic Acid, Diisoundecyl Ester (9CI), DIUP, Jayflex DIUP, 1,2-Benzenedicarboxylic Acid, 1,2-Diisoundecyl Ester
IUPAC-namn bis(9-methyldecyl) benzene-1,2-dicarboxylate
LEDER CC(C)CCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCCC(C)C
Specifikationer
Kemiskt namn eller material Diisoundecyl Phthalate (Mixture of Branched Isomers)
Kvantitet 1 g
InChI formel InChI=1S/C30H50O4/c1-25(2)19-13-9-5-7-11-17-23-33-29(31)27-21-15-16-22-28(27)30(32)34-24-18-12-8-6-10-14-20-26(3)4/h15-16,21-22,25-26H,5-14,17-20,23-24H2,1-4H3
Formel vikt 474.371