Läs mer
Tocris Bioscience™ Dilazep dihydrochloride
Beskrivning
Coronary and cerebral vasodilator, suppresses the effects of ischemia. Inhibitor of platelet aggregation and of membrane transport of nucleosides. Inhibits adenosine uptake.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dilazep dihydrochloride |
| CAS | 20153-98-4 |
| Kvantitet | 50 mg |
| Molekylformel | C31H46Cl2N2O10 |
| Formel vikt | 677.62 |
| Renhet | >99% |
| Molekylvikt (g/mol) | 677.61 |