Learn More
Trajan Scientific and Medical™ Diluter/Dispenser Syringes for PSD/3 and PSD/4 Pumps
Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for PSD/3 and PSD/4 Pumps has the knowledge and expertise that goes into manufacturing an SGE syringe.
Brand:  Trajan Scientific and Medical™ 005692
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15258483
  • 1075.00 SEK / Each
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Diluter/Dispenser Syringe for PSD/3 and PSD/4 Pumps
Specifications
Specifications
30 mm stroke, Luer Lock connection
125μL
For use with PSD/3 and PSD/4 pumps
Videos