Learn More
Trajan Scientific and Medical™ Diluter/Dispenser Syringes for PSD/3 and PSD/4 Pumps
Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for PSD/3 and PSD/4 Pumps has the knowledge and expertise that goes into manufacturing an SGE syringe.
Brand:  Trajan Scientific and Medical™ 005692
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15258483
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Diluter/Dispenser Syringe for PSD/3 and PSD/4 Pumps
Specifications
Specifications
30 mm stroke, Luer Lock connection
125μL
For use with PSD/3 and PSD/4 pumps
Videos