Learn More
Trajan Scientific and Medical™ Diluter/Dispenser Syringes for V3 Pumps
Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for V3 Pumps has the knowledge and expertise that goes into manufacturing an SGE syringe.
Brand:  Trajan Scientific and Medical™ 005693
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15268483
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Diluter/Dispenser Syringe for V3 Pumps
Specifications
Specifications
30mm stroke, 1/4-28 UNF thread connection, 4.8mm hole in plunger
100μL
For use with V3 pumps
Videos