Learn More
Trajan Scientific and Medical™ Diluter/Dispenser Syringes for V3 Pumps
Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for V3 Pumps has the knowledge and expertise that goes into manufacturing an SGE syringe.
Brand:  Trajan Scientific and Medical™ 005693
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15268483
  • 810.00 SEK / Each
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Diluter/Dispenser Syringe for V3 Pumps
Specifications
Specifications
30mm stroke, 1/4-28 UNF thread connection, 4.8mm hole in plunger
100μL
For use with V3 pumps
Videos