Learn More
Trajan Scientific and Medical™ Diluter/Dispenser Syringes for V3 Pumps
Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for V3 Pumps has the knowledge and expertise that goes into manufacturing an SGE syringe.
Brand:  Trajan Scientific and Medical™ 007693
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15278493
  • 1165.00 SEK / Each
Estimated Shipment: 17-07-2024
to see stock.
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Diluter/Dispenser Syringe for V3 Pumps
Specifications
Specifications
30mm stroke, 1/4-28 UNF thread connection, 4.8mm hole in plunger
500 μL
V3 Pump