Läs mer
Tocris Bioscience™ DIM-C-pPhOH
Beskrivning
DIM-C-pPhOH is a Nur77 (NR4A1) antagonist. Inhibits TGF-β induced cell migration of breast cancer cell lines. Promotes ROS/endoplasmic reticulum stress and proapoptotic pathways in pancreatic cancer cell lines. Mimics effects of Nur77 RNAi silencing.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 1,1-Bis(3'-indolyl)-1-(p-hydroxyphenyl)methane |
| CAS | 151358-47-3 |
| Analysprocentintervall | >98% |
| Kvantitet | 5 mg |
| Molekylformel | C23H18N2O |
| Formel vikt | Observed MW: 338.408 |