Läs mer
DIM-C-pPhOH, Tocris Bioscience™
Nur77 antagonist
Brand: Tocris Bioscience 6377/25
This item is not returnable.
View return policy
Description
DIM-C-pPhOH is a Nur77 (NR4A1) antagonist. Inhibits TGF-β induced cell migration of breast cancer cell lines. Promotes ROS/endoplasmic reticulum stress and proapoptotic pathways in pancreatic cancer cell lines. Mimics effects of Nur77 RNAi silencing.Specifications
| 1,1-Bis(3'-indolyl)-1-(p-hydroxyphenyl)methane | |
| 151358-47-3 | |
| >98% | |
| 25 mg |