Läs mer
Dimenthyl Succinate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dimenthyl Succinate |
| CAS | 34212-59-4 |
| Molekylformel | C24H42O4 |
| Kvantitet | 25 g |
| Synonym | 1,4-bis[(1R,2S,5R)-5-methyl-2-(1-methylethyl)cyclohexyl] Ester Butanedioic Acid, Bis[(1R,2S,5R)-5-methyl-2-(1-methylethyl)cyclohexyl] Ester Butanedioic Acid, [1R-[1alpha(1R*,2S*,5R*),2beta,5alpha]]-Bis[5-methyl-2-(1-methylethyl)cyclohexyl] Ester Butanedioic Acid, Succinate Menthol, (-)-Bis(l-menthyl) Succinate, (-)-Dimenthyl Succinate, Dimenthyl Succinate |
| InChI formel | InChI=1S/C24H42O4/c1-15(2)19-9-7-17(5)13-21(19)27-23(25)11-12-24(26)28-22-14-18(6)8-10-20(22)16(3)4/h15-22H,7-14H2,1-6H3/t17-,18-,19+,20+,21-,22-/m1/s1 |
| LEDER | CC(C)[C@@H]1CC[C@@H](C)C[C@H]1OC(=O)CCC(=O)O[C@@H]2C[C@H](C)CC[C@H]2C(C)C |
| IUPAC-namn | bis[(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] butanedioate |
| Molekylvikt (g/mol) | 394.59 |
| Formel vikt | 394.3083 |