missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Dimesitylborinic acid, 98%

Brand:  Alfa Aesar™ H55455.06

Additional Details : CAS Number : 20631-84-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15491698

  • 1012.36 SEK / 5g


Estimated Shipment date
from Supplier 07-05-2018
Add to basket

Specifications

Chemical Identifiers

Dimesitylborinic acid
266.19
Bis(2,4,6-trimethylphenyl)boranol
20631-84-9
MFCD01863507

Specifications

Dimesitylborinic acid
Solid
144° to 146°C
MFCD01863507
Bis(2,4,6-trimethylphenyl)boranol
20631-84-9
98%
266.19
5g
Videos
SDS