Learn More
Dimethallyl carbonate, 97%, Thermo Scientific Chemicals
425.24 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
It is used as an intermediate in enantioselective tsuji enol silane allylation reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an intermediate in enantioselective tsuji enol silane allylation reaction.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used as an intermediate in enantioselective tsuji enol silane allylation reaction.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Boiling Point | 199°C to 202°C |
Quantity | 5 g |
Formula Weight | 170.21 |
Percent Purity | 97% |
Chemical Name or Material | Dimethallyl carbonate |