Läs mer
Dimethisoquin Hydrochloride, LoGiCal
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dimethisoquin Hydrochloride |
| CAS | 2773-92-4 |
| Analyt- eller komponentnamn | Dimethisoquin Hydrochloride |
| Frakt skick | Room Temperature |
| Molekylformel | C17 H24 N2 O . Cl H |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | Quinisocaine Hydrochloride, Ethanamine, 2-[(3-butyl-1-isoquinolinyl)oxy]-N,N-dimethyl-, hydrochloride (1:1), Ethanamine, 2-[(3-butyl-1-isoquinolinyl)oxy]-N,N-dimethyl-, monohydrochloride (9CI), Isoquinoline, 3-butyl-1-[2-(dimethylamino)ethoxy]-, hydrochloride (6CI,7CI), Isoquinoline, 3-butyl-1-[2-(dimethylamino)ethoxy]-, monohydrochloride (8CI), 3-Butyl-1-(2-dimethylaminoethoxy)isoquinoline hydrochloride, Chinisocaine hydrochloride, Dimethisoquin hydrochloride, Isochinol, Pruralgin, Quinisocaine hydrochloride, Quotane, Quotane hydrochloride, 1-(beta-Dimethylaminoethoxy)-3-n-butylisoquinoline hydrochloride |
| InChI formel | InChI=1S/C17H24N2O.ClH/c1-4-5-9-15-13-14-8-6-7-10-16(14)17(18-15)20-12-11-19(2)3;/h6-8,10,13H,4-5,9,11-12H2,1-3H3;1H |
| Lösningstyp | Neat |
| LEDER | Cl.CCCCc1cc2ccccc2c(OCCN(C)C)n1 |
| Visa mer |