Läs mer
Dimethocaine Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dimethocaine (hydrochloride) |
| CAS | 553-63-9 |
| Molekylformel | C16 H26 N2 O2 . Cl H |
| Kvantitet | 10 mg |
| Synonym | 1-Propanol, 3-(diethylamino)-2,2-dimethyl-, 1-(4-aminobenzoate), hydrochloride (1:1), 1-Propanol, 3-(diethylamino)-2,2-dimethyl-, 4-aminobenzoate (ester), monohydrochloride (9CI), 1-Propanol, 3-(diethylamino)-2,2-dimethyl-, p-aminobenzoate (ester), monohydrochloride (8CI), 1-Aminobenzoyl-2,2-dimethyl-3-diethylaminopropanol hydrochloride, 3-Diethylamino-2,2-dimethylpropyl p-aminobenzoate hydrochloride, Diethylaminoneopentyl alcohol hydrochloride p-aminobenzoate, Dimethocaine hydrochloride, Larocaine hydrochloride |
| InChI formel | InChI=1S/C16H26N2O2.ClH/c1-5-18(6-2)11-16(3,4)12-20-15(19)13-7-9-14(17)10-8-13;/h7-10H,5-6,11-12,17H2,1-4H3;1H |
| LEDER | Cl.CCN(CC)CC(C)(C)COC(=O)c1ccc(N)cc1 |
| IUPAC-namn | [3-(diethylamino)-2,2-dimethylpropyl] 4-aminobenzoate;hydrochloride |
| Molekylvikt (g/mol) | 314.85 |
| Formel vikt | 314.1761 |