missing translation for 'onlineSavingsMsg'
Learn More

Dimethomorph-d8, TRC

Product Code. 30374655
Change view
Click to view available options
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30374655 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30374655 Supplier Toronto Research Chemicals Supplier No. TRCD46053225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Dimethomorph-d8
CAS 1346606-71-0
Molekylformel C21 D8 H14 Cl N O4
Kvantitet 25 mg
Synonym Dimethomorph D8 (Morpholine D8)
InChI formel InChI=1S/C21H22ClNO4/c1-25-19-8-5-16(13-20(19)26-2)18(15-3-6-17(22)7-4-15)14-21(24)23-9-11-27-12-10-23/h3-8,13-14H,9-12H2,1-2H3/b18-14+/i9D2,10D2,11D2,12D2
LEDER [2H]C1([2H])OC([2H])([2H])C([2H])([2H])N(C(=O)\C=C(/c2ccc(Cl)cc2)\c3ccc(OC)c(OC)c3)C1([2H])[2H]
IUPAC-namn (E)-3-(4-chlorophenyl)-3-(3,4-dimethoxyphenyl)-1-(2,2,3,3,5,5,6,6-octadeuteriomorpholin-4-yl)prop-2-en-1-one
Molekylvikt (g/mol) 395.906
Formel vikt 395.174
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.