Läs mer
Dimethyl 1,10-decanedicarboxylate, 98%, Thermo Scientific™
Produktkod. 11352696
Change view
Kvantitet:
10 g
50 g
Förpackningsstorlek:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11352696 10 g 10g
11362696 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11352696 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer L08757.09
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1731-79-9
Molekylformel C14H26O4
Molekylvikt (g/mol) 258.358
MDL-nummer MFCD00043632
InChI-nyckel IZMOTZDBVPMOFE-UHFFFAOYSA-N
Synonym dimethyl 1,10-decanedicarboxylate, dodecanedioic acid dimethyl ester, dodecanedioic acid, dimethyl ester, dimethyl 1,12-dodecanedioate, 1,12-dimethyl dodecanedioate, dodecanedioic acid dimethyl, dimethyl dodecane-1,12-dioate, dimethyldodecanedioate, dodecanedioic acid, 1,12-dimethyl ester, acmc-209e5w
PubChem CID 74415
IUPAC-namn dimethyl dodecanedioate
LEDER COC(=O)CCCCCCCCCCC(=O)OC
Specifikationer
Kemiskt namn eller material Dimethyl 1,10-decanedicarboxylate
Smältpunkt 27°C to 32°C
Kokpunkt 187°C to 188°C (14 mmHg)
Linjär formel CH3O2C(CH2)10CO2CH3
Kvantitet 10 g
Beilstein 1790424
Formel vikt 258.36
Procent renhet 98%