missing translation for 'onlineSavingsMsg'
Läs mer

Dimethyl 1,4-Dihydro-2,6-diisopropyl-4-(4-fluorophenyl)-pyridine-3,5-dicarboxylate, TRC

Produktkod. 30374137
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30374137 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Product Code. 30374137 Supplier Toronto Research Chemicals Supplier No. TRCD47171050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Dimethyl 1,4-Dihydro-2,6-diisopropyl-4-(4-fluorophenyl)-pyridine-3,5-dicarboxylate
CAS 132008-67-4
Molekylformel C21 H26 F N O4
Kvantitet 50 mg
InChI formel InChI=1S/C21H26FNO4/c1-11(2)18-16(20(24)26-5)15(13-7-9-14(22)10-8-13)17(21(25)27-6)19(23-18)12(3)4/h7-12,15,23H,1-6H3
LEDER COC(=O)C1=C(NC(=C(C1c2ccc(F)cc2)C(=O)OC)C(C)C)C(C)C
IUPAC-namn dimethyl 4-(4-fluorophenyl)-2,6-di(propan-2-yl)-1,4-dihydropyridine-3,5-dicarboxylate
Molekylvikt (g/mol) 375.43
Formel vikt 375.1846
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.