Läs mer
Dimethyl 3,3'-Dithiodipropionate, TRC
Produktkod. 30372577
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30372577 250 mg
1 options
Produktkod. 30372577 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD472715250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dimethyl 3,3'-Dithiodipropionate
CAS 15441-06-2
Molekylformel C8H14O4S2
Kvantitet 250 mg
Synonym Bis(methoxycarbonylethyl) Disulfide, 3,3'-Dithiodipropionic Acid Dimethyl Ester, 3,3'-Dithiodipropionic Acid Dimethyl Ester, NSC 638709
InChI formel InChI=1S/C8H14O4S2/c1-11-7(9)3-5-13-14-6-4-8(10)12-2/h3-6H2,1-2H3
LEDER COC(=O)CCSSCCC(=O)OC
Molekylvikt (g/mol) 238.32