missing translation for 'onlineSavingsMsg'
Läs mer

Dimethyl 3-(3-(Trifluoromethyl)phenoxy)-2-oxopropylphosphonate, TRC

Produktkod. 30351605
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30351605 1 g
30354586 250 mg
30358671 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30351605 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD1862501G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 54094-19-8
Molekylformel C12H14F3O5P
Molekylvikt (g/mol) 326.21
Synonym Dimethyl P-[2-oxo-3-[3-(trifluoromethyl)phenoxy]propyl]phosphonate, 1-Dimethoxyphosphoryl-3-[3-(trifluoromethyl)phenoxy]propan-2-one, Dimethyl [2-oxo-3-[3-(trifluoromethyl)phenoxy]propyl]phosphonate, Dimethyl [2-oxo-3-[[(alpha,alpha,alpha-trifluoro-m-tolyl)oxy]propyl]phosphonate, Dimethyl [3-((3-trifluoromethyl)phenoxy)-2-oxopropyl]phosphonate, [2-Oxo-3-[3-(trifluoromethyl)phenoxy]propyl]phosphonic Acid Dimethyl Ester
IUPAC-namn 1-dimethoxyphosphoryl-3-[3-(trifluoromethyl)phenoxy]propan-2-one
LEDER COP(=O)(CC(=O)COc1cccc(c1)C(F)(F)F)OC

Specifications

Kemiskt namn eller material Dimethyl 3-(3-(Trifluoromethyl)phenoxy)-2-oxopropylphosphonate
Kvantitet 1 g
InChI formel InChI=1S/C12H14F3O5P/c1-18-21(17,19-2)8-10(16)7-20-11-5-3-4-9(6-11)12(13,14)15/h3-6H,7-8H2,1-2H3
Formel vikt 326.0531
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.