Läs mer
Dimethyl 3-Hydroxyphthalate, TRC
Produktkod. 30369163
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30369163 10 mg
1 options
Produktkod. 30369163 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD47346010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dimethyl 3-Hydroxyphthalate
CAS 36669-02-0
Molekylformel C10H10O5
Kvantitet 10 mg
Synonym 3-Hydroxy-1,2-benzenedicarboxylic Acid Dimethyl Ester, 3-Hydroxyphthalic Acid Dimethyl Ester
InChI formel InChI=1S/C10H10O5/c1-14-9(12)6-4-3-5-7(11)8(6)10(13)15-2/h3-5,11H,1-2H3
LEDER COC(=O)c1cccc(O)c1C(=O)OC
Molekylvikt (g/mol) 210.18
Formel vikt 210.0528