Läs mer
Dimethyl 4-(2-Chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, TRC
Produktkod. 30133975
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30133975 2.5 g 2.5g
30007895 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133975 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4693502.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 43067-01-2
Molekylformel C17 H18 Cl N O4
Molekylvikt (g/mol) 335.78
Synonym Dimethyl 4-(2-Chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, Amlodipine Besilate Imp. G (EP)
IUPAC-namn dimethyl 4-(2-chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
LEDER COC(=O)C1=C(C)NC(=C(C1c2ccccc2Cl)C(=O)OC)C
Specifikationer
Kemiskt namn eller material dimethyl 4-(2-chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 g
InChI formel InChI=1S/C17H18ClNO4/c1-9-13(16(20)22-3)15(11-7-5-6-8-12(11)18)14(10(2)19-9)17(21)23-4/h5-8,15,19H,1-4H3
Formel vikt 335.0924
Procent renhet >95%