missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 4-chloropyridine-2,6-dicarboxylate, 98%, Thermo Scientific Chemicals

Catalog Number p-7056090
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
5371-70-0
C9H8ClNO4
229.616
MFCD02612781
NFXKYKHKNUFOKB-UHFFFAOYSA-N
dimethyl 4-chloro-2,6-pyridinedicarboxylate, methyl 4-chloropyridine-2,6-dicarboxylate, 2,6-dimethyl 4-chloropyridine-2,6-dicarboxylate, 2,6-pyridinedicarboxylic acid, 4-chloro-, dimethyl ester, 4-chloropyridine-2,6-dicarboxylic acid,dimethyl ester, methyl 4-chloro-6-methoxycarbonyl pyridine-2-carboxylate, 4-chloro-2,6-pyridinedicarboxylic acid dimethyl ester, pubchem20951, pych37, ksc496e3d
237845
dimethyl 4-chloropyridine-2,6-dicarboxylate
COC(=O)C1=CC(=CC(=N1)C(=O)OC)Cl
This item is not returnable. View return policy
5371-70-0
C9H8ClNO4
229.616
MFCD02612781
NFXKYKHKNUFOKB-UHFFFAOYSA-N
dimethyl 4-chloro-2,6-pyridinedicarboxylate, methyl 4-chloropyridine-2,6-dicarboxylate, 2,6-dimethyl 4-chloropyridine-2,6-dicarboxylate, 2,6-pyridinedicarboxylic acid, 4-chloro-, dimethyl ester, 4-chloropyridine-2,6-dicarboxylic acid,dimethyl ester, methyl 4-chloro-6-methoxycarbonyl pyridine-2-carboxylate, 4-chloro-2,6-pyridinedicarboxylic acid dimethyl ester, pubchem20951, pych37, ksc496e3d
237845
dimethyl 4-chloropyridine-2,6-dicarboxylate
COC(=O)C1=CC(=CC(=N1)C(=O)OC)Cl

Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 139°C to 144°C
Color Yellow
Quantity 250 mg
Beilstein 197075
Solubility Information Slightly soluble in water.
Formula Weight 229.62
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Dimethyl 4-chloropyridine-2,6-dicarboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'