Läs mer
Dimethyl 5-Iodoisophthalate, TRC
Produktkod. 30371088
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30371088 1 g
1 options
Produktkod. 30371088 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4741801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dimethyl 5-Iodoisophthalate
CAS 51839-15-7
Molekylformel C10H9IO4
Kvantitet 1 g
Synonym 3-(Methoxycarbonyl)-5-iodobenzoic Acid Methyl Ester, Dimethyl 5-Iodoisophthalate
InChI formel InChI=1S/C10H9IO4/c1-14-9(12)6-3-7(10(13)15-2)5-8(11)4-6/h3-5H,1-2H3
LEDER O=C(OC)C1=CC(I)=CC(C(OC)=O)=C1
IUPAC-namn dimethyl 5-iodoisophthalate
Molekylvikt (g/mol) 320.08
Formel vikt 319.95